Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS139550

Pyrex® round media storage bottles and reusable screw caps

capacity 50 mL

Synonym(s):

Pyrex® media bottles, glass media bottles, laboratory bottles, plastic bottle, plastic bottles, round bottles, storage bottle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43
Pricing and availability is not currently available.

material

borosilicate glass bottle
clear bottle
cylindrical bottle
narrow-mouth bottle
orange polypropylene cap (plug seal)

sterility

non-sterile

feature

autoclavable
cap
graduations
reusable

packaging

case of 10

manufacturer/tradename

Corning 1395-50

availability

not available in EU

capacity

50 mL

diam. × H

46 mm × 88 mm

fitting

GL 32 neck thread

Looking for similar products? Visit Product Comparison Guide

General description

Pyrex heavy-duty round media storage bottles can be used for storage as well as for mixing and sampling. These media storage bottles Include linerless, one-piece, autoclavable, orange PP plug-seal caps with drip-free pouring rings. Optional red PBTP caps and red ETFE pouring rings are available separately, for dry heat sterilization (180°C).

Pyrex round media storage bottles are made of borosilicate glass with permanent, white enamel graduations and marking spots.

Caution: Bottles larger than 2 L should NOT be used with bottle-top filter units or in other applications involving vacuum/pressure, as breakage may occur. DO NOT tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage.

A wide range of optional caps are available for these bottles:
  • Vented membrane cap with 0.22 μm PTFE hydrophobic membrane
  • Red PBT (polybutylene terephthalate) solid caps and ETFE pouring rings for dry heat sterilization (180°C)
  • PBT caps with silicone septa for syringe sampling or introduction of reagents
These heavy duty 50 mL PYREX round bottles can be used for storage as well as mixing and sampling. They include a linerless, one-piece autoclavable orange polypropylene plug seal GL32 threaded cap and a drip-free pouring ring. Optional red PBT caps and red ETFE pouring rings are available for dry heat sterilization (180°C). Bottles have permanent white enamel graduations and marking spots.

Caution: DO NOT tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage.

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service