Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

707.fl

99072807, mouse spleen, Lymphoblastoid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

707.fl, 99072807

biological source

mouse spleen

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblastoid

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse friend Leukaemia

Cell Line Description

Has a high ability to differentiate into erythroblastoid cells.

Culture Medium

RPMI 1640 + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9 x 10,000 cells/ml 37°C 5% CO2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service