Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATDC5

99072806, mouse unknown/unspecified, Epithelial-like

Synonym(s):

ATDC-5 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

ATDC5, 99072806

biological source

mouse unknown/unspecified

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse 129 teratocarcinoma AT805 derived

Cell Line Description

The chondrogenic cell line ATDC5 has been derived from teratocarcinoma AT805. Cells showed a sequential transition of phenotype in vitro. They encompass stages from mesenchymal condensation to calcification. Bone morphogenetic protein-2 (BMP-2) stimulated the sequential progression of early- and late-phase differentiation. Activation of parathyroid hormone (PTH)/PTH-related peptide (PTHrP) receptor resulted in inhibition of this differentiation capacity.

Application

ATDC5 has been used to study cartilage differentiation.

Culture Medium

DMEM: Ham′s F12 (1:1) + 2mM Glutamine + 5% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) i.e. seeding at 1-3 x 10,000 cells/cm² using 0.25% trypsin or trypsin/EDTA;CO2; 37°C subculture every 2-3 days.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service