Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F5-5.F1

99072803, mouse spleen, Lymphoblastoid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

F5-5.F1, 99072803

biological source

mouse spleen

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblastoid

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Lymphocyte, friend leukemia

Cell Line Description

Investigation of differentiation-inducible leukaemia cells is considered important in the understanding of leukaemic or neoplastic state. Several ascites lines derived from friend virus-induced erythoid leukemia show an erythroid morphology and also, unusually high haem synthesis. Most cultured lines of friend virus-induced leukemia in mice, are known to be inducible to differentiate towards erythocytes. F-5-5.F1 is susceptible to the induction by both dimethyl and butyric acid.

Culture Medium

RPMI 1640 + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:10 i.e. seeding at 1x10,000 cells/cm2 using 0.25% trypsin or

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service