Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COR-L23/R23-

96042337, human unknown/unspecified, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

COR-L23/R23-, 96042337

biological source

human unknown/unspecified

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian lung large cell carcinoma,drug-resistant revertant

Cell Line Description

The revertant lung cancer cell line COR-L23/R23- was generated by growing the doxorubicin-selected, resistant variant COR-L23/R (Sigma Catalogue number. 96042339) without drug exposure for 24-28 weeks. The cell line, also known as COR-L23/Rev, overexpresses multidrug resistance -associated protein (MRP), but to a lesser extent than the parent line. It was shown that the cell line has the ability to recover quickly, similar levels of MRP expression and resistance as COR-L23/R after a transient exposure to the MDR-drugs doxorubicin and vincristine.

Application

Studies on lung cancer, drug-resistance analysis

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11
D13S317: 10,11
D16S539: 11,13
D5S818: 10,12
D7S820: 9
THO1: 9
TPOX: 8,11
vWA: 19

Culture Medium

RPMI 1640 +2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service