Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NB4 1A3

89121405, mouse nerve, Neuroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

NB4 1A3, 89121405

biological source

mouse nerve

growth mode

Adherent

karyotype

Unstable

morphology

Neuroblast

products

Acetylcholinesterase, choline acetylase, tyrosine hydroxylase

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse C-1300 Neuroblastoma

Cell Line Description

Cloned from the C1300 mouse neuroblastoma line. Adapted to culture by alternate in vivo and in vitro passage. Cells have characteristics of mature neurons and secrete the enzymes for the synthesis of the neurotransmitters, choline acetylase and tyrosine hydroxylase. Cells are susceptible to VSV, HSV and Vaccinia viruses.

Application

Tumourigenicity, enzymatic and virus studies: VSV, HSV, vaccinia virus

Culture Medium

Ham′s F10 + 2mM Glutamine + 15% Horse Serum (HS) + 2.5% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service