Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HR5-CL11

95091411, human cervix, Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
human cervix
growth mode:
Adherent
karyotype:
Not specified
morphology:
Epithelial-like
products:
Not specified
receptors:
Not specified
relevant disease(s):
cancer

Product Name

HR5-CL11, 95091411

biological source

human cervix

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human cervix carcinoma, HeLa derivative, transformed

Cell Line Description

HR5-CL11 cells were obtained after transfection of HeLa cells with plasmids coding for a transcriptional transactivator and transferring G418 and hygromycin resistance. This tetracycline-controlled transactivator (a fusion between the tet repressor and the activating domain of the herpes simplex virus VP16 transcription activation domain) can stimulate the expression of a reporter gene up to 100,000 fold. Subsequently these cells were stably transfected with a luciferase reporter unit. Luciferase expression in the HR5-CL11 cells is highly stimulated by doxycycline.

Application

Gene expression and activation studies

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 to 1:20 i.e. seeding at 2x1,000 to 2x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service