Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

X1/5

95051229, human cervix, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

X1/5, 95051229

biological source

human cervix

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human (HeLa) with tTA

Cell Line Description

X1/5 is a HeLa cell line stably transfected with a tet repressor fused with the activating domain of virion protein 16 of herpes simplex virus thus generating a tetracycline-controlled transactivator (tTA) This stimulates transcription from a minimal promoter sequence derived from the human cytomegalovirus promoter IE combined with tet operator sequences. Integration of a luciferase gene controlled by a tTA-dependent promoter into this tTA producing HeLa cell line leads to high levels of tetracycline-sensitive luciferase expression. The luciferase activity can be regulated over up to five orders of magnitude depending on the concentration of tetracycline in the culture medium (0-1ug/ml). Specific and stringent control of the activity of an individual gene (luciferase) in the mammalian cell can be monitored. It is also suitable for creation of on/off situations and analysis of mRNA decay rates under physiological conditions

Application

Specific and stringent control of the activity of an individual gene (luciferase) in a mammalian cell can be monitored. It is also suitable for creation of

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 1% Vitamins + 500μg/ml G418 + 300μg/ml Hygromycin + 10% Foetal Bovine Serum (FBS). Freeze cells in 40% medium, 10% Dimethyl Sulphoxide (DMSO) and 50% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 to 1:20 i.e. seeding at 5x1,000 to 2x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA. Alternatively cells can be washed with PBS and subcultured after addition of PBS with 0.8mM EDTA (30% of original volume)

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service