Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PNT1A human

95012614, human prostate, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

PNT1A human, 95012614, Normal prostate epithelium imortalized with SV40.

biological source

human prostate

growth mode

Adherent

karyotype

Aneuploid

morphology

Epithelial

products

Not specified

receptors

Dihydrotestosterone receptor

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

Cell Line Origin

Human post pubertal prostate normal, immortalised with SV40

Cell Line Description

Established by immortalisation of normal adult prostatic epithelial cells by transfection with a plasmid containing SV40 genome with a defective replication origin. The primary culture was obtained from the prostate of a 35 year old male at post mortem. The cells contain the SV40 genome and express large T protein. They present the phenotype of differentiated luminal prostatic cells with the expression of cytokeratin 8 and 18 (markers for luminal glandular epithelia) and vimentin. Cytokeratin14, a marker of epithelial basal cells, is not expressed. PNT1A cells are non-tumourigenic in nude mice.

Application

PNT1A has been used to determine the variation in metabolite levels of human DU145 prostate cancer and PNT1A cells after treatment with selenomethionine (SeM) and Se-methylselenocysteine (SeMSC). It has also been used to study the effects of angiotensin II (Ang II) and relaxin 2 (RLN2) on the viability, proliferation, adhesion, migration and invasion of PNT1A cells.
Study of proliferation, oncogene activation and differentiation of adult prostatic cells and initial steps leading to transformation.

Culture Medium

RPMI 1640 + 2mM Glutamine + 6 -10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:10 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2;37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service