Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B16 melanoma 4A5 Cell Line from mouse

94042254, mouse skin, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

B16 melanoma 4A5 Cell Line from mouse, Mouse melanoma, producing melanin, 94042254

biological source

mouse skin

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast-like

products

Melanin

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse melanoma, melanin producing

Cell Line Description

Derived from a melanoma from the skin of a C57BL/6 strain mouse, showing fibroblast-like characteristics, which produces melanin. Cells may lose ability to produce pigmentation in long term culture.

Application

B16 melanoma 4A5 cell line has been used to study the spectral and biological characteristics of copper (II) mesoporphyrinic complexes. It has also been used to evaluate the cytotoxicity of JaZ-30, a matrix metalloproteinase-2 (MMP-2) inhibitor.

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service