Skip to Content
MilliporeSigma
All Photos(2)

Documents

106-05A

Sigma-Aldrich

Human Dermal Fibroblasts: HDF, adult

Synonym(s):

HDF cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

human neonatal foreskin or adult skin (normal)

Quality Level

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 46

morphology

Fibroblast

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Dermal Fibroblasts are responsible for producing the extracellular matrix forming the connective tissue of the skin, and play a crucial role during wound healing. HDF provide an excellent model system to study many aspects of cell physiology, and have been utilized in dozens of research publications, particularly those related to skin biology and reprogramming/induced pluripotency studies. HDF were used to create, along with human synovyocytes, induced pluripotent stem cells (iPSC) by using the now classic “Yamanaka cocktail” of Oct3/4, Sox2, Klf4, and c-Myc, the discovery for which Dr. Shinya Yamanaka was awarded the Nobel Prize in 2012 (Takanashi, 2007; 2009), and to characterize iPSC general properties, miRNA expression profiles and differentiation potential (Hayashi, 2010; Ohta, 2011; Koyanagi-Aoi, 2013; Noguchi, 20130; Razak, 2013; Sakurai, 2013; Tanaka, 2013; Yanagida, 2013)

Cell Line Origin

Skin

Application

injury recovery, connective tissue research, laminin and fibronectin production, extracellular matrix, tissue organization, wound healing, fibroblasts growth factor response

Components

Fibroblast Basal Medium containing 10% FBS & 10% DMSO.

Preparation Note

  • Primary culture, >500,000 cells in Fibroblast Basal Medium containing 10% FBS & 10% DMSO.
  • Can be cultured at least 16 doublings.

Culture Medium

Fibroblast Growth Medium (116-500) all-in-one ready to use media

Subculture Routine

Please refer to the HDF Culture Protocol.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

required but not provided

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michele Galluccio et al.
Molecular biology reports, 47(9), 7283-7289 (2020-08-11)
It is well established that Escherichia coli represents a powerful tool for the over-expression of human proteins for structure/function studies. In many cases, such as for membrane transporters, the bacterial toxicity or the aggregation of the target protein hamper the
Manthan Patel et al.
BMC genetics, 21(1), 84-84 (2020-07-31)
The human CGGBP1 binds to GC-rich regions and interspersed repeats, maintains homeostasis of stochastic cytosine methylation and determines DNA-binding of CTCF. Interdependence between regulation of cytosine methylation and CTCF occupancy by CGGBP1 remains unknown. By analyzing methylated DNA-sequencing data obtained
Lucie Peterková et al.
Tissue & cell, 58, 121-129 (2019-05-28)
Surface modification is an important step in making a synthetic polymer cytocompatible. We have previously reported improved cytocompatibility of immortalized human keratinocytes (HaCaT) with the otherwise bioinert fluorinated ethylene propylene (FEP) upon treatment with argon plasma discharge. In this article

Protocols

This protocol contains: Storage - Preparation for Culturing - Culturing HDF - Subculturing HDF

This protocol contains: Storage - Preparation for Culturing - Culturing HDF - Subculturing HDF

This protocol contains: Storage - Preparation for Culturing - Culturing HDF - Subculturing HDF

This protocol contains: Storage - Preparation for Culturing - Culturing HDF - Subculturing HDF

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service