Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R2C

89031606, rat testis, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

R2C, 89031606

biological source

rat testis

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Steroid hormones; 20-hydroxypregn-4-en-3-one; progesterone

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat testis Leydig cell tumour

Cell Line Description

A rat testis Leydig cell tumour cell line derived from a 2-month-old male WFu rat. Virus susceptibility: herpes simplex; vaccinia; pseudorabies; vesicular stomatitis (Indiana). Resistant to poliovirus 2. Stimulation by other hormones or cAMP is not required for hormone secretion.

Culture Medium

HAMS F10 + 2mM Glutamine + 15% Horse serum + 2.5% Foetal Bovine Serum (FBS). Do not use heat-inactivated serum.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:3 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin/EDTA;5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service