Skip to Content
MilliporeSigma

AB9698

Anti-Neuron Specific Enolase (NSE) Antibody

Chemicon®, from chicken

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IHC, WB
Citations:
2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

chicken

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human, rat

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ENO2(2026)

Immunogen

Synthetic peptides from human/rat/mouse Neuron Specific Enolase.

Application

Anti-Neuron Specific Enolase (NSE) Antibody is an antibody against Neuron Specific Enolase (NSE) for use in IC, IH & WB.
Immunohistochemistry: 1:50-1:100 using 2% paraformaldehyde fixed tissues.
Immunocytochemistry: 1:50-1:100 using 2% paraformaldehyde fixed cells.
Western blotting: On mouse brain extracts, blot shows a single band at 47 kDa.
Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers

Biochem/physiol Actions

Recognizes Neuron Specific Enolase (NSE).

Physical form

Affinity purified chicken IgY. Two different antibodies were combined to make this product. Liquid in PBS, pH 7.2 with 0.02% sodium azide.

Preparation Note

Maintain at 2-8°C in undiluted aliquots for up to 6 months after date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: AB5619

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library