Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ARH 77

88121201, human blood, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

ARH 77, 88121201

biological source

human blood

growth mode

Suspension

karyotype

Hypodiploid

morphology

Lymphoblast

products

Immunoglobulin G (IgG1) (kappa)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human plasma cell leukaemia

Cell Line Description

Derived from a 33 year old female with plasma cell leukaemia. The cells synthesise IgG1 (kappa-light chain) and are EBNA positive.Although the cells are reported to originate from a patient with a plasma cell leukaemia , it has now been shown that this is in fact an EBV transformed B lymphoblastoid cell line .

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 6,10
D13S317: 11,13
D16S539: 9,13
D5S818: 10,13
D7S820: 7,12
THO1: 8,9.3
TPOX: 8
vWA: 17

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 1-4x100,000 cells/ml; 5% CO2; 37°C. Maximum cell density 1-2x1,000,000 cells/ml.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service