Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HeLa S3

87110901, human cervix, Epithelial

Synonym(s):

HeLa-S3 Cells, HeLa/S3 Cells, HeLaS3 Cells, S3 HeLa Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
human cervix
growth mode:
Suspension
karyotype:
Modal no. 68
morphology:
Epithelial
products:
Not specified
receptors:
Not specified
relevant disease(s):
cancer

Product Name

HeLa S3, 87110901

biological source

human cervix

growth mode

Suspension

karyotype

Modal no. 68

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human cervix carcinoma

Cell Line Description

Derived from the parent HeLa cell line. The cells have been adapted to grow in suspension (spinner) culture and have the same virus susceptibility as the parent line. Very useful in the clonal analysis of mammalian cell populations relating to chromosomal variation, cell nutrition and plaque forming ability. The cells should be handled under laboratory containment level 2.

Application

HeLa S3 has been used to study the cytotoxicity of dinuclear titanium-salan complexes. It has also been used to perform in vitro genotoxicity tests (comet and micronucleus) to study the effects of occupational exposure to carcinogens on sewage workers.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 9,10
D13S317: 14
D16S539: 9,10
D5S818: 11,12
D7S820: 8,12
THO1: 7
TPOX: 8,12
vWA: 16,18

Culture Medium

SMEM + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) +10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Culture in a spinner flask. This cell line may adhere if the culture is not agitated.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service