Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Ma-Mel-19

13012460, human skin, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

Ma-Mel-19, from human skin, 13012460

biological source

human skin

description

Human melanoma, metastasis

growth mode

Adherent

morphology

Epithelial

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Description

Ma-Mel-19 (ESTDAB-152) was established from the cutaneous/subcutaneous metastasis in a 62 year-old female patient with metastatic melanoma (superficial spreading melanoma stage IV). The Ma-Mel-19 cell line is one of more than 170 characterized melanoma cell lines collected during the ESTDAB project (European Searchable Tumour Line Database) part of the European Commission′s fifth framework infrastructures programme (contract no. QLRI-CT-2001-01325). ESTDAB contains information on HLA genotype and surface expression of HLA molecules, expression of tumour antigens, antigen processing capability, production of and response to cytokines and chemokines, apoptosis regulation and expression of adhesion/accessory molecules. Such information allows researchers to identify cells possessing specific parameters important for studies of immunity, immunogenetics, gene expression, metastasis, response to chemotherapy, and other tumour biological experimentation.

DNA Profile

STR-PCR Data:

Amelogenin: X
CSF1PO: 10,12
D13S317: 8,12
D16S539: 11,13
D5S818: 9,12
D7S820: 8,9
THO1: 9.3
TPOX: 8
vWA: 14,18

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Fetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) seed new cultures at 1-3 x10,000 cells/cm² using 0.05% trypsin or trypsin/EDTA; 5% CO2; 37°C. Saturation density approx 8 x10,000 cells/cm² .

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service