Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

293 GTP-AC-free

5011003, human kidney (embryonic), Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

293 GTP-AC-free, 05011003

biological source

human kidney (embryonic)

growth mode

Aggregates in suspension

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Embryo Kidney, serum-free

Cell Line Description

The human embryo kidney cell line 293 (Sigma Catalogue number. 85120602) adapted to grow in the animal component free medium Gene Therapy Medium 3 (Sigma G9916). This cell line grows in clumps in suspension. The original 293 cell line from which this cell line was derived was transformed with sheared human Ad5 DNA.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11,12
D13S317: 12
D16S539: 9,13
D5S818: 8,9
D7S820: 11,12
THO1: 7,9.3
TPOX: 11
vWA: 16,19

Culture Medium

Gene Therapy Medium 3 (Sigma G9916) + 2mM Glutamine. When freezing cells down use 50:50 fresh culture medium:conditioned medium plus 10% DMSO.

Subculture Routine

Viability may be poor on resuscitation and may initially decrease further. Full recovery may take up to 2 weeks. A centrifugation step to remove the cryoprotectant is essential. Rapidly thaw the frozen ampoule in a water bath at 37°C for 1-2 minutes. Tran

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service