Key Documents
EP5895200001
Rotor for Eppendorf® 5910 R Centrifuge
S-4xUniversal, incl. 4 universal buckets, pkg of 4 ea
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 4.0 mL to 1,000 mL tubes and flasks and plates
S-4xUniversal, incl. 4 universal buckets
packaging
pack of 1 ea
pkg of 4 ea
manufacturer/tradename
Eppendorf® 5895200001
parameter
4,500 max. RCF (4,347 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5910R
General description
Rotor for Eppendorf(R)5910R Centrifuge
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)