Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CHO/dhFr-AC-free cell line

5011002, hamster ovary, Suspension

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

CHO/dhFr-AC-free cell line, 05011002

biological source

hamster ovary

growth mode

Aggregates in suspension

karyotype

Not specified

morphology

Suspension

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Hamster Chinese Ovary, serum-free

Cell Line Description

This is a derviative of the CHO/dhFr- cell line, Sigma Catalogue number. 94060607 which has been adapted to grow in the animal component free medium CHO DHFR (Sigma C8862). The CHO/dhFr- cell line lacks the enzyme dihydrofolate reductase (DHFR) which is necessary for purine synthesis. In the absence of exogenous purines, this enzyme is required for growth. Hence the CHO/dhFr- cell line has growth requirements for hypoxanthine (or adenine), glycine and thymidine. However, the cells will continue to grow without the addition of hypoxanthine and thymidine because the cells can obtain sufficient from the medium. α MEM minus does not contain the ribonucleosides or the deoxyribonucleosides, and will not support the growth of CHO/dhFr- or its serum free adaptation as neither contains the gene to produce DHFR.

Culture Medium

CHO DHR medium (Sigma C8862) + 2mM Glutamine + 100uM Hypoxanthine + 16uM Thymidine (e.g. use HT supplement Sigma H0137). Addition of Aminopterin (Sigma A5159) 0.4uM is also advised. When freezing cells use freeze medium composed of 50:50 fresh:conditioned medium + 10% DMSO.

Subculture Routine

Maintain cultures between approximately 3-9x105 cells/ml; 5% CO2; 37°C. Cells grow as aggregates in suspension. Viability may be poor on resuscitation and may initially decrease further. Full recovery may take up to 2 weeks.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service