Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

C9995

Sigma-Aldrich

Anti-Cullin-4B antibody produced in rabbit

enhanced validation

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Cul4B, Anti-MRXHF2, Anti-MRXSC, Anti-SFM2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~100 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.0 mg/mL

technique(s)

western blot: 0.7-1.5 μg/mL using HEK-293T cells expressing human Cullin-4B.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CUL4B(8450)
mouse ... Cul4b(72584)
rat ... Cul4b(302502)

General description

Mammalian cells express two distinct but highly homologous cullin-4 proteins, cullin-4A and cullin-4B. Cul4A and Cul4B form individual E3 ligases. In contrast to Cul4A, Cul4B has been shown to be localized to the nucleus.

Application

Anti-Cullin-4B antibody produced in rabbit is suitable for western blotting at a working concentration of 0.7-1.5 μg/mL using HEK-293T cells expressing human Cullin-4B. The antibody was used for immunoblotting in a study to demonstrate that CUL4B E3 ubiquitin ligase plays a key role in targeting Jab1 (Jun activation domain-binding protein) for degradation and regulates bone morphogenetic protein signaling.

Biochem/physiol Actions

Cullin-4 (Cul4) proteins are the core components of ubiquitin E3 ligases that regulate cell cycle, genome stability and DNA replication. Cul4B functions as an E3 ligase involved in selecting specific targets for ubiquitylation. It associates with damage-specific DNA binding proteins DDB1 and DDB2 (UV-damaged DNA) and functions as a substrate-recruiting factor for Cul4B E3 ligases. Cul4B ubiquitylates several target proteins including β-catenin, cyclin E, and histone H2A Mutations in this gene has been associated with x-linked mental retardation.
Cullin-4B facilitates the transfer of damaged-DNA binding protein 1 (DDB1) into the nucleus independently of DDB2. In contrast to cullin-4A (Cul4A), cullin-4B (Cul4B) does not ubiquitinate DDB2.

Target description

Cullin-4B forms a complex that functions as an E3 ubiquitin ligase and catalyzes the polyubiquitination of specific protein substrates in the cell. The protein interacts with a ring finger protein, and is required for the proteolysis of several regulators

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pengyu Li et al.
Oncology letters, 14(2), 1265-1274 (2017-08-16)
Cullin 4B (Cul4B), a scaffold protein that assembles the ubiquitin ligase complex, is involved in a wide variety of physiological and developmental processes, such as cell cycle progression, DNA damage response and gene expression regulation. Cul4B is overexpressed in various
The Xeroderma Pigmentosum Group E Gene Product DDB2 Is a Specific Target of Cullin 4A in Mammalian Cells
Nag A, et al.
Molecular and Cellular Biology, 21(20), 6738-6747 (2001)
Rachana Tripathi et al.
Journal of biosciences, 32(6), 1133-1138 (2007-10-24)
Beta-catenin is the key transducer of Wingless-type MMTV integration site family member (Wnt) signalling, upregulation of which is the cause of cancer of the colon and other tissues. In the absence of Wnt signals, beta-catenin is targeted to ubiquitin-proteasome-mediated degradation.
Yanjun Li et al.
Oncogenesis, 9(2), 20-20 (2020-02-15)
Given that colorectal cancer stem cells (CCSCs) play key roles in the tumor dormancy, metastasis, and relapse, targeting CCSCs is a promising strategy in cancer therapy. Here, we aimed to identify the new regulators of CCSCs and found that Cullin
Yongxin Zou et al.
American journal of human genetics, 80(3), 561-566 (2007-02-03)
We reevaluated a previously reported family with an X-linked mental retardation syndrome and attempted to identify the underlying genetic defect. Screening of candidate genes in a 10-Mb region on Xq25 implicated CUL4B as the causative gene. CUL4B encodes a scaffold

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service