MilliporeSigma
Cul4B is a novel prognostic marker in cholangiocarcinoma.
Oncology letters (2017-08-16)
Pengyu Li, Lili Zhang, Muyi Yang, Mei Qi, Xing Jin, Bo Han
ABSTRACT
Cullin 4B (Cul4B), a scaffold protein that assembles the ubiquitin ligase complex, is involved in a wide variety of physiological and developmental processes, such as cell cycle progression, DNA damage response and gene expression regulation. Cul4B is overexpressed in various solid tumors. However, the prognostic value and role of Cul4B in cholangiocarcinoma (CCA) is largely unknown. The present study demonstrated that Cul4B was overexpressed in 21 (26.6%) of 79 patients with intrahepatic CCA, and in 40 (28.6%) of 140 patients with extrahepatic CCA (EHCC). Kaplan-Meier survival analysis suggested that Cul4B expression is an unfavorable prognostic factor in EHCC patients. Notably, Cul4B and epidermal growth factor receptor expression define a subset of CCA patients with poor prognosis.
Sigma-Aldrich
Anti-Cullin-4B antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution