Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C9868

Sigma-Aldrich

Anti-CRTH2 (GPR44) antibody produced in chicken

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~43 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: 1:500-1:1,000

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... GPR44(11251)
mouse ... Gpr44(14764)
rat ... Gpr44(309212)

Immunogen

synthetic peptide corresponding to residues 15-33 of human CRTH2 (GRP44).

Target description

CRTH2 (chemoattractant receptor-homologousmolecule), which is also known as the putative Gprotein-coupled receptor (GPR44), is expressed onTh2 cells and is a novel member of the G proteincoupledleukocyte chemoattractant receptor family,which is

Physical form

solution in phosphate buffered saline, containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service