Skip to Content
MilliporeSigma

C0867

Anti-CRTH2 (GRP44) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Chemoattractant Receptor-homologous Molecule expressed on TH2 Cells, Anti-Putative G-Protein Coupled Receptor 44

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
3
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~43 kDa by calculation

species reactivity

rat, mouse, human

technique(s)

western blot: 1:500-1:1,000 using CRTH2-transfected Jurkat cells

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... GPR44(11251)
mouse ... Gpr44(14764)
rat ... Gpr44(309212)

Immunogen

peptide corresponding to the human CRTH2 (GRP44) protein, near the C-terminus (amino acids 329-348).

Application

Anti-CRTH2 (GRP44) antibody produced in rabbit is suitable for western blotting at a working dilution of 1:500-1:1,000 using CRTH2- transfected Jurkat cells.

Biochem/physiol Actions

CRTH2 (chemoattractant receptor-homologous molecule), is known as the putative G protein-coupled receptor (GP44), is expressed on T helper 2 (Th2) cells. It is also referred as DP2, DL1R and CD294. It′s a human protein and belongs to G protein-coupled leukocyte chemo attractant receptor family. It is not expressed on human amniocytes or myocytes. On activation of CRTH2, PGD(2) induces human osteoclast apoptosis and the apoptosis intrinsic pathway. CRTH2, CD4 and T cells all together may be involved in the pathogenesis of IgG4-related Lacrimal gland enlargement (LGE) causing Mikulicz′s disease (MD).

Physical form

Solution of 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library