Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C8749

Sigma-Aldrich

Copeptin trifluoroacetate human

≥95% (HPLC)

Synonym(s):

H-Ala-Ser-Asp-Arg-Ser-Asn-Ala-Thr-Gln-Leu-Asp-Gly-Pro-Ala-Gly-Ala-Leu-Leu-Leu-Arg-Leu-Val-Gln-Leu-Ala-Gly-Ala-Pro-Glu-Pro-Phe-Glu-Pro-Ala-Gln-Pro-Asp-Ala-Tyr-OH trifluoroacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C177H279N49O58 · xC2HF3O2
CAS Number:
Molecular Weight:
4021.40 (free base basis)
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

biological source

human

Assay

≥95% (HPLC)

form

film or powder

color

colorless to light yellow

solubility

H2O: >1 mg/mL

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CC(C)C[C@H](NC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](C)NC(=O)CNC(=O)[C@H](C)NC(=O)[C@@H]1CCCN1C(=O)CNC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H](NC(=O)[C@H](C)NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CO)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CO)NC(=O)[C@H](C)N)[C@@H](C)O)C(C)C)C(=O)N[C@@H](C)C(=O)NCC(=O)N[C@@H](C)C(=O)N2CCC[C@H]2C(=O)N[C@@H](CCC(O)=O)C(=O)N3CCC[C@H]3C(=O)N[C@@H](Cc4ccccc4)C(=O)N[C@@H](CCC(O)=O)C(=O)N5CCC[C@H]5C(=O)N[C@@H](C)C(=O)N[C@@H](CCC(N)=O)C(=O)N6CCC[C@H]6C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](C)C(=O)N[C@@H](Cc7ccc(O)cc7)C(O)=O

InChI

1S/C177H279N49O58.C2HF3O2/c1-81(2)64-107(152(260)194-89(16)141(249)190-77-131(236)193-95(22)171(279)223-60-30-40-123(223)166(274)205-106(51-56-134(240)241)174(282)226-63-32-41-124(226)167(275)215-113(70-97-34-25-24-26-35-97)159(267)204-105(50-55-133(238)239)173(281)224-61-31-39-122(224)165(273)198-93(20)144(252)203-104(49-54-128(181)233)172(280)225-62-33-42-125(225)168(276)216-116(74-136(244)245)154(262)195-92(19)145(253)217-118(175(283)284)71-98-43-45-99(230)46-44-98)207-150(258)102(47-52-126(179)231)201-169(277)138(87(13)14)220-161(269)112(69-86(11)12)209-148(256)100(36-27-57-187-176(183)184)199-155(263)109(66-83(5)6)210-157(265)111(68-85(9)10)211-156(264)108(65-82(3)4)206-143(251)91(18)192-130(235)76-189-142(250)90(17)197-164(272)121-38-29-59-222(121)132(237)78-191-147(255)115(73-135(242)243)213-158(266)110(67-84(7)8)208-151(259)103(48-53-127(180)232)202-170(278)139(96(23)229)221-146(254)94(21)196-153(261)114(72-129(182)234)212-163(271)120(80-228)219-149(257)101(37-28-58-188-177(185)186)200-160(268)117(75-137(246)247)214-162(270)119(79-227)218-140(248)88(15)178;3-2(4,5)1(6)7/h24-26,34-35,43-46,81-96,100-125,138-139,227-230H,27-33,36-42,47-80,178H2,1-23H3,(H2,179,231)(H2,180,232)(H2,181,233)(H2,182,234)(H,189,250)(H,190,249)(H,191,255)(H,192,235)(H,193,236)(H,194,260)(H,195,262)(H,196,261)(H,197,272)(H,198,273)(H,199,263)(H,200,268)(H,201,277)(H,202,278)(H,203,252)(H,204,267)(H,205,274)(H,206,251)(H,207,258)(H,208,259)(H,209,256)(H,210,265)(H,211,264)(H,212,271)(H,213,266)(H,214,270)(H,215,275)(H,216,276)(H,217,253)(H,218,248)(H,219,257)(H,220,269)(H,221,254)(H,238,239)(H,240,241)(H,242,243)(H,244,245)(H,246,247)(H,283,284)(H4,183,184,187)(H4,185,186,188);(H,6,7)/t88-,89-,90-,91-,92-,93-,94-,95-,96+,100-,101-,102-,103-,104-,105-,106-,107-,108-,109-,110-,111-,112-,113-,114-,115-,116-,117-,118-,119-,120-,121-,122-,123-,124-,125-,138-,139-;/m0./s1

InChI key

PBRJDIQIICROHQ-URAAVIGWSA-N

Biochem/physiol Actions

Copeptin, the C-terminal part of the vasopressin prohormone is a stable and sensitive surrogate marker for AVP release.
Copeptin, the C-terminal part of the vasopressin prohormone is a stable and sensitive surrogate marker for AVP release. Copeptin, a 39-amino acid glycopeptide that comprises the C-terminal part of the Arginine vasopressin (AVP) precursor (CT-proAVP), is cosynthesized with AVP. AVP is a primary secretagogue of the hypothalamo–pituitary–adrenal axis, a key hormone for maintaining fluid balance and vascular tone, and is a marker of acute endogenous stress. However, it has a short half-life and measurement of mature AVP is difficult and subject to errors. Copeptin in contrast is stable and easily measured.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service