Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C4701

Sigma-Aldrich

Vertical mini-gel Comb

10 cm wide, for use with 1.5 mm x 10 wells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105305
NACRES:
NC.01

product name

Combs for vertical mini-gel, 10 cm wide, size 1.5 mm × 10 wells

size

1.5 mm × 10 wells

Quality Level

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service