Skip to Content
MilliporeSigma
All Photos(1)

Documents

C2671

Sigma-Aldrich

Corticotropin Releasing Factor bovine

≥90% (HPLC), Synthetic

Synonym(s):

CRF, CRH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C206H340N60O63S
CAS Number:
Molecular Weight:
4697.33
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

Quality Level

Assay

≥90% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

Gene Information

cow ... CRH(280755)

Amino Acid Sequence

Ser-Gln-Glu-Pro-Pro-Ile-Ser-Leu-Asp-Leu-Thr-Phe-His-Leu-Leu-Arg-Glu-Val-Leu-Glu-Met-Thr-Lys-Ala-Asp-Gln-Leu-Ala-Gln-Gln-Ala-His-Asn-Asn-Arg-Lys-Leu-Leu-Asp-Ile-Ala

Biochem/physiol Actions

CRF is a hypothalamic peptide that releases ACTH and endorphin from the anterior pituitary. It is also a neurotransmitter in the central nervous system, and is involved in autonomic and endocrine responses to stress.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacob M Maskal et al.
Scientific reports, 11(1), 22527-22527 (2021-11-20)
In utero heat stress alters postnatal physiological and behavioral stress responses in pigs. However, the mechanisms underlying these alterations have not been determined. The study objective was to characterize the postnatal hypothalamic-pituitary-adrenal axis response of in utero heat-stressed pigs. Pigs
James Koh et al.
Journal of cellular and molecular medicine, 20(2), 351-359 (2015-12-08)
Primary hyperparathyroidism (PHPT) is a common endocrine neoplastic disorder caused by a failure of calcium sensing secondary to tumour development in one or more of the parathyroid glands. Parathyroid adenomas are comprised of distinct cellular subpopulations of variable clonal status
Mitchell Stewart Fleming et al.
Frontiers in endocrinology, 11, 603538-603538 (2020-12-18)
We recently characterized two paralogs of the thyrotropin (TSH) beta subunit in Atlantic salmon, tshβa and tshβb, issued from teleost-specific whole genome duplication. The transcript expression of tshβb, but not of tshβa, peaks at the time of smoltification, which revealed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service