Skip to Content
MilliporeSigma
All Photos(1)

Documents

SRP6269

Sigma-Aldrich

MMP-1 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

CLG, CLGN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (N-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

calculated mol wt 52.7 kDa
observed mol wt 52-55 kDa (DTT-reduced. Phe 20 is the predicted N-terminus.)

packaging

pkg of 10 and 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MMP-1(4312)

General description

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMPs are secreted as inactive pro-proteins which are activated when cleaved by extracellular proteinases. The enzyme encoded by this gene degrades type IV and V collagens. Studies in rhesus monkeys suggest that the enzyme is involved in IL-8-induced mobilization of hematopoietic progenitor cells from bone marrow, and murine studies suggest a role in tumor-associated tissue remodeling. Thrombospondins, intervertebral disc proteins, regulate the effective levels of matrix metalloproteinases (MMPs) 2 and 9, which are key effectors of ECM remodeling. Matrix metalloproteinase-1 (MMP-1) also known as interstitial collagenase and fibroblast collagenase. MMP1 is expressed by fibroblasts, keratinocytes, endothelial cells, monocytes and macrophages. MMP-1 breaks down the interstitial collagens, types I, II, and III. MMP1 can degrade a broad range of substrates including types I, II, III, VII, VIII, and X collagens as well as casein, gelatin, myelin basic protein, L Selectin, proTNF, IL1β, IGFBP3, IGFBP5, pro MMP2 and pro MMP9.

Physical form

Lyophilized from 0.22 μm filtered solution in MES, NaCl, CaCl2 and Brij­35. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jayeeta Giri et al.
Cell reports, 30(6), 1923-1934 (2020-02-13)
Mesenchymal stromal cell (MSC)-based therapy for inflammatory diseases involves paracrine and efferocytotic activation of immunosuppressive interleukin-10+ (IL-10+) macrophages. The paracrine pathway for MSC-mediated IL-10+ macrophage functionality and response to tissue injury is not fully understood. In our present study, clodronate pre-treatment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service