Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-14009

Sigma-Aldrich

Human Umbilical Vein Endothelial Cells 2 (HUVEC 2)

Pooled, isolated in Growth Medium 2, 1 million cells per pellet

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

application(s)

cell analysis (DNA, RNA, and protein)

shipped in

dry ice

storage temp.

−20°C

Application

Primary Human Umbilical Vein Endothelial Cells (HUVEC) are isolated from the vein of the umbilical cord. They are commonly used for physiological and pharmacological investigations, such as macromolecule transport, blood coagulation, angiogenesis, and fibrinolysis. HUVEC are isolated from pooled donors (from up to four different umbilical cords) in Endothelial Cell Growth Medium 2, which is free of ECGS and contains additional IGF and VEGF. (Human Umbilical Artery Endothelial Cells (HUAEC) from the same donor are available on request.)

Quality

Rigid quality control tests are performed for each lot of the corresponding cells. They are tested for cell viability, cell morphology, growth performance and for cell-type specific markers. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).

Warning

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.

Physical form

Pellet is dissolved in RNAlater® for downstream analysis of DNA, RNA or protein profiles. Cells are not revivable.

Subculture Routine

Click here for more information.

Other Notes

The Cell Pellets can be stored indefinitely at -20°C, and for up to 1 month at 4°C. Note: The Cell Pellets will not freeze at -20°C. Always centrifuge product briefly before opening vial.

Legal Information

RNAlater is a registered trademark of Ambion, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Primary cells from PromoCell are available in pelleted form for research applications like gene expression analysis, and cell/protein characterization experiments.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service