Skip to Content
MilliporeSigma
All Photos(1)

Documents

C-12915

Sigma-Aldrich

Human M2 Macrophages (hMDM-MCSF(-))

Monocyte-derived, single donor, 1.5 million cryopreserved cells

Synonym(s):

hMDM-MCSF(-) cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

human blood (monocyte)

packaging

pkg of 1,500,000 cells

morphology

(macrophages)

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Cell Line Origin

Monocyte

Application

Cryopreserved human monocyte-derived macrophages (hMDM) are convenient and easy-to-handle. The thawed cells plate into all tissue culture vessel formats and can be maintained as adherent, biologically functional cultures for several weeks. Optionally, user-customizable activation of the cells can be performed.Cryopreserved human macrophages are produced from monocytes in M2 Macrophage Generation Media DXF and are available as non-activated, fully qualified M2- (M-CSF) polarized cells. After 9-10 days of differentiation, the M2 macrophages are cryopreserved using Cryo-SFM. Each cryo vial contains more than 1.5 million viable cells after thawing.

Warning

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.

Subculture Routine

Click here for more information.

Other Notes

Recommended Plating Density: 200000 cells per cm2Tested Markers: CD163 positive, CD68 positiveNote: The plated cells do not proliferate. Recommended plating density for obtaining an adherent cell monolayer with 70-90% confluency.

Recommended products

Recommended Primary Cell Culture Media:Link
Recommended Primary Cell Culture Media:Link

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Yeast plasmid selection: Utilizes auxotrophic yeast strains for plasmid selection, enabling growth on synthetic media for better yield.

Yeast plasmid selection: Utilizes auxotrophic yeast strains for plasmid selection, enabling growth on synthetic media for better yield.

Overview of primary cells and media usage in biomedical culture applications highlights differences from cell lines.

Overview of primary cells and media usage in biomedical culture applications highlights differences from cell lines.

See All

Protocols

Generate phagocytic M1 and M2 macrophages from human PBMCs in 10 days in serum-free and xeno-free cell culture media. Explore over 350 PromoCell products.

Generate phagocytic M1 and M2 macrophages from human PBMCs in 10 days in serum-free and xeno-free cell culture media. Explore over 350 PromoCell products.

Generate phagocytic M1 and M2 macrophages from human PBMCs in 10 days in serum-free and xeno-free cell culture media. Explore over 350 PromoCell products.

Generate phagocytic M1 and M2 macrophages from human PBMCs in 10 days in serum-free and xeno-free cell culture media. Explore over 350 PromoCell products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service