Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

C1052

Sigma-Aldrich

Cellulose Azure

cellulase substrate

Synonym(s):

Cellulose-azure

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.83

form

powder

Looking for similar products? Visit Product Comparison Guide

General description

Cellulose azure is an insoluble cellodextrin together with water soluble reactive dye.

Application

Cellulose Azure has been used as a substrate for determining cellulase activity. It has also been used as a supplement in Modified Melin-Norkrans (MMN) medium for determining the lignocellulolytic activity of the fungi.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel root-fungus symbiosis in Ericaceae: sheathed ericoid mycorrhiza formed by a hitherto undescribed basidiomycete with affinities to Trechisporales
Vohnik M, et al.
Testing, 7(6), e39524-e39524 (2012)
Martin Vohník et al.
PloS one, 7(6), e39524-e39524 (2012-07-05)
Ericaceae (the heath family) are widely distributed calcifuges inhabiting soils with inherently poor nutrient status. Ericaceae overcome nutrient limitation through symbiosis with ericoid mycorrhizal (ErM) fungi that mobilize nutrients complexed in recalcitrant organic matter. At present, recognized ErM fungi include
Quantification of cellulase activity using cellulose-azure
Lai TE, et al.
Talanta, 69(1), 68-72 (2006)
Pulsed laser deposition and characterization of cellulase thin films
Cicco N, et al.
Applied Surface Science, 278, 223-225 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service