Skip to Content
MilliporeSigma
All Photos(1)

Documents

BUA444-EP

Buckets for Eppendorf® A-4-44 Rotor

(2 x 50 mL conical tubes), pack of 2

Synonym(s):

Lid for Eppendorf® FA-45-48-11 Rotor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103910

feature

Holds 2 x 50 mLcentrifuge tubes

packaging

pack of 2

manufacturer/tradename

Eppendorf® 022637452 (NA)
Eppendorf® 5804706005 (EU)

technique(s)

centrifugation: suitable

capacity

(2 x 50 mL conical tubes)

compatibility

for use with Rotor A-4-44

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service