About This Item
UNSPSC Code:
41103910
Manufacturer/tradename:
Eppendorf® 022638866 (NA), Eppendorf® 5810742005 (EU)
Feature:
Holds x flex bucket, MTP/Flex bucket
feature
Holds x flex bucket, MTP/Flex bucket
packaging
pack of 2
manufacturer/tradename
Eppendorf® 022638866 (NA), Eppendorf® 5810742005 (EU)
technique(s)
centrifugation: suitable
General description
MTP/Flex buckets are designed for use with Eppendorf® A-4-81 rotor. These buckets have flat bottoms with rubber mats. MTP/Flex buckets supplied as a set of 2 can accommodate 4 microplates(MTP), 2 cell culture plates, or 1 deep well plate(DWP) per bucket.
Application
MTP/Flex bucket is suitable for A-4-81 rotor for 5910 Ri centrifuge.
Features and Benefits
- Designed for Eppendorf® A-4-81 and S-4x500 rotors
- Ideal for use with IsoRack and cell culture flask adapters as well as microplates(MTP), and deep well plates (DWP)
- MTP/Flex buckets with a maximum speed of 4,000 RPM and a maximum RCF of 2,900 x g are ideal for high-speed centrifugation
- MTP/Flex buckets have a 163 mm centrifugation radius
Legal Information
Eppendorf is a registered trademark of Eppendorf SE
Still not finding the right product?
Explore all of our products under Buckets for Eppendorf® A-4-81 Rotor
Choose from one of the most recent versions:
Already Own This Product?