Key Documents
BMSW1105131000
Sigma-Aldrich
Accuris M1 Grade Calibration Weight Set
pack of 13 pieces, size 1-1000 g, stainless steel
About This Item
UNSPSC Code:
41100000
NACRES:
NB.71
material
stainless steel
packaging
pack of 13 pieces
manufacturer/tradename
Benchmark Scientific W1105-13100
size
1-1000 g
General description
Basic, compact balances with readability in the range of 0.1g to 0.01g can be calibrated with M1 Class weights.
Each individual weight and weight set from Accuris is supplied with an individual Certificate of Verification confirming that the weight(s) meet OIML regulations and tolerances for the specific weight class.
The economy M1 Calibration Weight Set includes 13 pieces: 1 x 1g, 2x2g, 1x5g, 1x10g, 2x20g, 1x50g, 1x100g, 2x200g, 1x500g and 1x1Kg. A protective carrying case is included.
Legal Information
Accuris is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)