Key Documents
BMSIPD96004M
Sigma-Aldrich
Benchmark Stainless Steel Bulk Beads
bead diam. 4 mm, acid washed
Synonym(s):
Stainless Steel Beads
About This Item
UNSPSC Code:
41100000
NACRES:
NA.82
material
stainless steel
packaging
pack of 5000 ea
manufacturer/tradename
Benchmark Scientific IPD9600-4M
bead diam.
4 mm
compatibility
stainless steel grinding jars
General description
Beads are alcohol washed and heat treated to remove contaminants and render them nuclease and protease free