Key Documents
BMSIPD960025S
Sigma-Aldrich
BeadBlaster 96 Grinding Jars for Hard Samples
stainless steel, capacity 25 mL
About This Item
UNSPSC Code:
41100000
NACRES:
NA.82
material
stainless steel
packaging
pack of 2
manufacturer/tradename
Benchmark Scientific IPD9600-25S
capacity
25 mL
compatibility
BeadBlaster 96
General description
Different materials have different grinding requirements. Very hard samples require a jar and grinding balls that are just as hard or harder for effective sample preparation. The jars feature threaded closures for leakproof performance with both solids and liquids. Stainless steel is a very hard material, and when combined with the stainless steel balls, provides for grinding of hard samples such as minerals, ores, polymers and circuit boards. Cryogrinding and also be performed with stainless steel jars and balls. These jars are available in 25ml and 50ml volumes with 10mm and 25mm grinding balls.
Application
Homogenization of cryo samples
Legal Information
BeadBlaster is a trademark of Benchmark Scientific, Inc.