Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BAH369042006

Bel-Art® Sterileware Scoop

Bent Handle, capacity 4 oz

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122403
NACRES:
NB.35

material

natural
polystyrene

packaging

package of 10

manufacturer/tradename

Bel-Art H36904-2006

L × W × H

191 mm × 50 mm × 42 mm

capacity

4 oz

Looking for similar products? Visit Product Comparison Guide

General description

The ergonomic, curved handle design of the Sterileware Bent Handle Scoops elevates the hand from the sample when compared to straight handle scoops. This reduces the possibility of contamination and exposure of hands or personal protective equipment (PPE) to hazardous substances while minimizing wrist strain.

Features and Benefits

Curved handle helps prevent incidental contact with sample source material

Flat bottom keeps scoop upright when placed on a flat surface; can be used as a weigh boat

Made from high-impact, lightweight, FDA grade polystyrene

Individually packaged and ready for one time use and disposal

Legal Information

Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service