Documents
B8157
Sigma-Aldrich
Nalgene® bottles, style 2006
capacity 60 mL
Synonym(s):
nalgene bottle, nalgene bottles
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
polypropylene cap
polypropylene copolymer bottle (PPCO)
round translucent bottle
Joint
Joints threaded neck (20)
description
Autoclavable
feature
with cap
packaging
pack of 12 ea
manufacturer/tradename
Nalgene® 2006-0002
bottle diam. × H
37 mm × 85 mm
capacity
60 mL
General description
Boston style (round) narrow-mouth bottles. Same profile as style 2002.
Features and Benefits
  • Nalgene® narrow-mouth bottles are ideal for long-term storage.
  • Made from polypropylene copolymer (PPCO) material for excellent sample clarity.
  • Highly chemical resistant to most acids, bases, and alcohols.
  • Autoclavable - with and without contents.
  • Laboratory-grade plastic materials for dependably low leachables and extractables.
  • Liner less caps ensures leak-proof seal for sample protection.
  • Light-weight - enables easy lifting and transporting.
  • Avoid UV exposure.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Visit the Document Library
Customers Also Viewed
Related Content
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.