Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYTGL200RD57RS

Gel cutting tips

O.D. 0.57 mm, round, racked, size 200 μL, pkg of 2x200tips/cs, sterile

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
O.D.:
0.57 mm
size:
200 μL
sterility:
sterile
feature:
DNA free
RNase free
barrier: no
endotoxin free
filter: no
packaging:
pkg of 2x200tips/cs
manufacturer/tradename:
Corning TGL-200RD-57-R-S

material

clear

sterility

sterile

feature

DNA free
RNase free
barrier: no
endotoxin free
filter: no

manufacturer/tradename

Corning TGL-200RD-57-R-S

packaging

pkg of 2x200tips/cs

O.D.

0.57 mm

size

200 μL

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service