AXYST150SSX
Screw cap tubes without caps, conical bottom, self-standing
size 1.5 mL, amber, pkg of 8x500caps/cs
Synonym(s):
centrifuge tubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
Recommended Products
material
conical bottom tube (standing)
polypropylene
sterility
non-sterile
feature
graduations: no
manufacturer/tradename
Axygen Inc ST-150-SS-X
Corning
packaging
pkg of 8x500caps/cs
size
1.5 mL
color
amber
Looking for similar products? Visit Product Comparison Guide
Related Categories
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service