Key Documents
AXYRESMW12HP
Reservoir
12 channel, high profile multiwells, size 7 mL, 25 ea of
Synonym(s):
storage reservoir
About This Item
UNSPSC Code:
41121522
NACRES:
NB.22
manufacturer/tradename
Axygen Inc RES-MW12-HP
Corning
packaging
25 ea of
size
7 mL
General description
  • Axygen® reagent reservoir is a multiple-well, high-profile reservoir manufactured using chemically resistant polypropylene.
  • Ideal for both manual and automated applications.
  • Designed with standard ANSI/SLAS footprint dimensions for microplates.
  • Features 12-Channel trough - extended length of the bottom surface.
  • Designed for 12-channel applications.
Legal Information
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)