Key Documents
AXYPCRAS600
Aluminum sealing film for thermal and general sealing
size 50 μm, pkg of 5x100sheets/cs
Synonym(s):
96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate
About This Item
UNSPSC Code:
41122410
NACRES:
NB.22
material
aluminum
sterility
non-sterile
manufacturer/tradename
Axygen Inc PCR-AS-600
Corning
packaging
pkg of 5x100sheets/cs
size
50 μm
General description
  • Axygen® aluminum sealing films are ideal for light-sensitive samples.
  • Uniform adhesive enables efficient well sealing.
  • Versatile sealing film can be used in various applications including PCR, Real-Time PCR to ELISA and cell culture.
  • Compatible with a wide range of reagents and temperatures.
Legal Information
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.