Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYPCR96LPABC

PCR microplate for ABI

low profile, size 96 wells, pkg of 4x25plates/cs, clear

Synonym(s):

96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
conical bottom wells
size:
96 wells
well volume:
100 μL

material

conical bottom wells

sterility

non-sterile

feature

lid: no
skirt (half skirt)

manufacturer/tradename

Axygen Inc PCR-96-LP-AB-C
Corning

packaging

pkg of 4x25plates/cs

size

96 wells

well volume

100 μL

color

clear

suitability

suitable for (RTPCR)

Looking for similar products? Visit Product Comparison Guide

General description

  • Axygen® 96-well semi-skirted PCR microplate is ideal for ABI Fast PCR 7500, 7900 9800, Veriti and StepOne systems.
  • Uniform ultra-thin wall thickness facilitates optimal thermal transfer.
  • Free of DNase-/RNase and nonpyrogenic.
  • Can be sealed with 8/12 strip caps, Axymat, or sealing film.
  • Low profile design optimized for fast PCR.
Products are available in China only.

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service