Key Documents
AXYP96450VCS
V-bottom assay plate
size 96 wells, V-bottom clear polypropylene wells, maximum volume 500 μL, pkg of 10x5plates/cs, sterile
Synonym(s):
V-Bottom Plate
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material:
V-bottom clear polypropylene wells
clear plate
size:
96 wells
material
V-bottom clear polypropylene wells
clear plate
sterility
sterile
feature
lid: no
skirt
manufacturer/tradename
Axygen Inc P-96-450V-C-S
Corning
packaging
pkg of 10x5plates/cs
maximum volume
500 μL
size
96 wells
working volume
450 μL
suitability
suitable for PCR
binding type
non-treated surface
General description
  • Axygen® V-bottom assay plates are manufactured using chemically resistant polypropylene.
  • The plates can withstand temperatures down to -80°C.
  • Ideal for sample collection, in vitro growth chambers, and long-term storage.
  • The ultra-flat surface of the plates can be sealed using heat sealing film for sample protection.
Legal Information
Axygen is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)