Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV44176

Sigma-Aldrich

Anti-SLC26A5 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-DFNB61, Anti-MGC118886, Anti-MGC118887, Anti-MGC118888, Anti-MGC118889, Anti-PRES, Anti-Solute carrier family 26, member 5 (prestin)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

81 kDa

species reactivity

horse, guinea pig, dog, rabbit, bovine, rat, mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Synthetic peptide directed towards the middle region of human SLC26A5

Application

Anti-SLC26A5 antibody produced in rabbit is suitable for western blotting at a concentration of 2.5μg/ml and for immunohistochemistry of paraffin-embedded tissue sections at a concentration of 4-8μg/ml.

Biochem/physiol Actions

SLC26A5 (Prestin), a member of SLC26 family, is an anion transporter that functions at microsecond rates. It is expressed at the basolateral membrane of cochlear outer hair cells and modulates the sensitivity of mammalian hearing. Mutations in the gene encoding for prestin result in neurosensory deafness.

Sequence

Synthetic peptide located within the following region: FSVTISMAKTLANKHGYQVDGNQELIALGLCNSIGSLFQTFSISCSLSRS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dmitry Gorbunov et al.
Nature communications, 5, 3622-3622 (2014-04-09)
Prestin (SLC26A5) is a member of the SLC26/SulP anion transporter family. Its unique quasi-piezoelectric mechanical activity generates fast cellular motility of cochlear outer hair cells, a key process underlying active amplification in the mammalian ear. Despite its established physiological role
Dalian Ding et al.
Frontiers in cell and developmental biology, 9, 643709-643709 (2021-06-11)
2-Hyroxypropyl-beta-cyclodextrin (HPβCD) is being used to treat Niemann-Pick C1, a fatal neurodegenerative disease caused by abnormal cholesterol metabolism. HPβCD slows disease progression, but unfortunately causes severe, rapid onset hearing loss by destroying the outer hair cells (OHC). HPβCD-induced damage is
Seth L Alper et al.
Molecular aspects of medicine, 34(2-3), 494-515 (2013-03-20)
The phylogenetically ancient SLC26 gene family encodes multifunctional anion exchangers and anion channels transporting a broad range of substrates, including Cl(-), HCO3(-), sulfate, oxalate, I(-), and formate. SLC26 polypeptides are characterized by N-terminal cytoplasmic domains, 10-14 hydrophobic transmembrane spans, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service