MilliporeSigma
  • Molecular architecture and the structural basis for anion interaction in prestin and SLC26 transporters.
Molecular architecture and the structural basis for anion interaction in prestin and SLC26 transporters.
Nature communications (2014-04-09)
Dmitry Gorbunov, Mattia Sturlese, Florian Nies, Murielle Kluge, Massimo Bellanda, Roberto Battistutta, Dominik Oliver
ABSTRACT
Prestin (SLC26A5) is a member of the SLC26/SulP anion transporter family. Its unique quasi-piezoelectric mechanical activity generates fast cellular motility of cochlear outer hair cells, a key process underlying active amplification in the mammalian ear. Despite its established physiological role, it is essentially unknown how prestin can generate mechanical force, since structural information on SLC26/SulP proteins is lacking. Here we derive a structural model of prestin and related transporters by combining homology modelling, MD simulations and cysteine accessibility scanning. Prestin's transmembrane core region is organized in a 7+7 inverted repeat architecture. The model suggests a central cavity as the substrate-binding site located midway of the anion permeation pathway, which is supported by experimental solute accessibility and mutational analysis. Anion binding to this site also controls the electromotile activity of prestin. The combined structural and functional data provide a framework for understanding electromotility and anion transport by SLC26 transporters.