Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV43810

Sigma-Aldrich

Anti-SLC38A1 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-ATA1, Anti-NAT2, Anti-SAT1, Anti-SNAT1, Anti-Solute carrier family 38, member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

54 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC38A1(81539)

General description

Solute carrier family 38, member 1/sodium-coupled neutral amino acid transporter 1 (SLC38A1, ATA1, NAT2, SAT1, SNAT1), which is expressed during embryogenesis, is a sodium-dependent transporter of neutral zwitterionic amino acids, such a glutamine. SLC38A1/SAT1 is believed to be involved in translocation of glutamine into GABAergic neurons to facilitate inhibitory neurotransmitter generation.

Specificity

Anti-SLC38A1 (AB1) polyclonal antibody reacts with canine and human solute carrier family 38, member 1 proteins.

Immunogen

Synthetic peptide directed towards the middle region of human SLC38A1

Application

Anti-SLC38A1 (AB1) polyclonal antibody is used to tag solute carrier family 38, member 1/sodium-coupled neutral amino acid transporter for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of solute carrier family 38, member 1 in transport of important neutral zwitterionic amino acids, such a glutamine, during embryogenesis and in neural function.

Biochem/physiol Actions

Amino acid transporters play essential roles in the uptake of nutrients, production of energy, chemical metabolism, detoxification, and neurotransmitter cycling. SLC38A1 is an important transporter of glutamine, an intermediate in the detoxification of ammonia and the production of urea. Glutamine serves as a precursor for the synaptic transmitter, glutamate.

Sequence

Synthetic peptide located within the following region: LLKNLGYLGYTSGFSLSCMVFFLIVVIYKKFQIPCIVPELNSTISANSTN

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O R Vaughan et al.
Journal of molecular endocrinology, 63(4), 239-248 (2019-09-11)
Excess maternal glucocorticoids reduce placental amino acid transport and fetal growth, but whether these effects are mediated directly on the syncytiotrophoblast remains unknown. We hypothesised that glucocorticoids inhibit mechanistic target of rapamycin (mTOR) signaling and insulin-stimulated System A amino acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service