Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV38543

Sigma-Aldrich

Anti-NRF1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-ALPHA-PAL, Anti-EWG, Anti-Nuclear respiratory factor 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

56 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NRF1(4899)

General description

Nuclear respiratory factor 1 (NRF1) is a transcription factor that activates cassettes of metabolic genes that regulate cellular respiratory and oxidative stress genes involved in heme biosynthesis, mitochondrial DNA transcription and replication and antioxidant enzymes. NRF1 and NRF2 function to mediate biogenomic coordination between nuclear and mitochondrial genomes.

The previously assigned protein identifier Q96AN2 has been merged into Q16656. Full details can be found on the UniProt database.

Specificity

Anti-NRF1 polyclonal antibody reacts with human nuclear respiratory factor 1.

Immunogen

Synthetic peptide directed towards the C terminal region of human NRF1

Application

Anti-NRF1 polyclonal antibody is used to tag nuclear respiratory factor 1 protein for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of nuclear respiratory factor 1 in respiratory and antioxidation processes and nuclear, mitochondrial biogenomic coordination.

Biochem/physiol Actions

NRF1 is a phosphorylated nuclear protein with a bZIP domain. This protein homodimerizes and functions as a transcription factor that activates the expression of some key metabolic genes regulating cellular growth and nuclear genes required for respiration, heme biosynthesis, and mitochondrial DNA transcription and replication. The protein has also been associated with the regulation of neurite outgrowth.This gene encodes a phosphorylated nuclear protein with a bZIP domain. This protein homodimerizes and functions as a transcription factor that activates the expression of some key metabolic genes regulating cellular growth and nuclear genes required for respiration, heme biosynthesis, and mitochondrial DNA transcription and replication. The protein has also been associated with the regulation of neurite outgrowth. Alternate transcriptional splice variants, which encode the same protein, have been characterized. Additional variants encoding different protein isoforms have been described but they have not been fully characterized.

Sequence

Synthetic peptide located within the following region: IVLSGETAAAVGALTGVQDANGLFMADRAGRKWILTDKATGLVQIPVSMY

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service