Skip to Content
MilliporeSigma
All Photos(2)

Documents

94829

Sigma-Aldrich

Poly(L-lactide)

average Mn 40,000

Synonym(s):

PLA, Poly(L-lactic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15O(C6H8O4)nH
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

description

ester terminated

Quality Level

form

solid

mol wt

average Mn 40,000

degradation timeframe

>3 years

Application

medical device

Other Notes

end group: hexadecyl ester

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Interest in utilizing biodegradable polymers for biomedical applications has grown since the 1960s.

AliAliphatic polyesters, including polylactide and polyglycolide, are biodegradable polymers widely used in medical applications.

AliAliphatic polyesters, including polylactide and polyglycolide, are biodegradable polymers widely used in medical applications.

AliAliphatic polyesters, including polylactide and polyglycolide, are biodegradable polymers widely used in medical applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service