Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV34053

Sigma-Aldrich

Anti-RGS20 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Regulator of G-protein signalling 20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

25 kDa

species reactivity

guinea pig, human, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RGS20(8601)

Immunogen

Synthetic peptide directed towards the middle region of human RGS20

Biochem/physiol Actions

Regulator of G protein signaling (RGS) proteins are regulatory and structural components of G protein-coupled receptor complexes. RGS proteins are GTPase-activating proteins for Gi class G-alpha proteins. They accelerate transit through the cycle of GTP binding and hydrolysis and thereby accelerate signaling kinetics and termination.

Sequence

Synthetic peptide located within the following region: NAFREFLRTEFSEENMLFWMACEELKKEANKNIIEEKARIIYEDYISILS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gang Li et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 117, 109112-109112 (2019-06-19)
Bladder cancer is a complicated disease with high rate of morbidity and mortality, in which proliferation and migration are both well acknowledged as aggressive phenotypes of bladder cancer cells. A better understanding of the mechanisms of tumor proliferation and migration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service