Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4500379

Sigma-Aldrich

Anti-TSH2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

OVC10-2, Ovarian cancer-related protein 10-2, Zinc finger protein 218

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 115 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TSHZ2(128553)

General description

Anti-TSH2 Antibody detects endogenous levels of total TSH2 protein.
TSH2/TSHZ2 (teashirt zinc finger homeobox 2) is a nuclear protein. It has four zinc-finger (ZF) domains, a homeobox domain and a coiled-coil domain. It is located on human chromosome 20q13.2.

Immunogen

The antiserum was produced against synthesized peptide derived from human TSH2.

Immunogen Range: 811-860

Biochem/physiol Actions

TSH2/TSHZ2 (teashirt zinc finger homeobox 2) acts as a C-terminal binding protein (CtBP)-dependent transcriptional repressor. At the time of mammary tumorigenesis, low level expression of TSH2 liberates GLI1 (glioma-associated oncogene homolog 1) from the nuclear repressor complex to update its transcriptional activity.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Down-regulation of the zinc-finger homeobox protein TSHZ2 releases GLI1 from the nuclear repressor complex to restore its transcriptional activity during mammary tumorigenesis
Miho Riku
Oncotarget, 7(5), 5690-5701 (2016)
Lessons learned from the initial sequencing of the pig genome: comparative analysis of an 8 Mb region of pig chromosome 17
Hart EA, et al.
Genome Biology (2007)
Madeline A Lancaster et al.
Nature, 501(7467), 373-379 (2013-09-03)
The complexity of the human brain has made it difficult to study many brain disorders in model organisms, highlighting the need for an in vitro model of human brain development. Here we have developed a human pluripotent stem cell-derived three-dimensional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service